NeuLog Electrocardiogram (EKG) Logger Sensor
Supplier: Wards
NUL218
470137-404EA 228.9 USD
470137-404
NeuLog Electrocardiogram (EKG) Logger Sensor
Data Loggers
This sensor measures the electrical signals released during muscle contractions.
  • For Experiments between One Second and 31 Days
  • Reusable Electrodes
  • Sensor Collects Data from Activity in Heart, Arm, Leg, or Jaw
  • For Measuring Electronic Signals Released During Muscle Contractions
The electrodes are reusable and can collect data from activity in the heart, arm, leg, or jaw.
It supports experiments lasting from one second to 31 days.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
Caution: For educational use only.