NeuLog Galvanic Skin Response (GSR) Logger Sensor
Supplier: Wards
NUL217
470137-402EA 185.15 USD
470137-402
NeuLog Galvanic Skin Response (GSR) Logger Sensor
Data Loggers
Measures the Psycho-Galvanic Reflex.
  • Max. sample rate 100 S/sec.
The Galvanic Skin Response (GSR) logger sensor measures the conductivity of our skin as it changes according to unconscious emotion effects such as sudden noise, smell, touch, pain or view. This sensor has two ranges: conductivity in micro Siemens and arbitrary numbers.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.