NeuLog Spirometer Logger Sensor
Supplier: Wards
NUL216
470137-400EA 222.3 USD
470137-400
NeuLog Spirometer Logger Sensor
Data Loggers
Track airflow and lung volume.
  • Maximum sample rate of 100 S/s
The spirometer enables human respiratory measurements. The sensor measures the air flow passing through and the volume (in liters) is calculated by the software. This sensor supports experiments lasting between 1 second and 31 days. For each probe you will need one or more of the following items: Battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WiFi module), WiFi module (not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Accessories information: Accessory item, replacement paper tubes, measure 10×6 cm.
Ordering information: The following items are required but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module).
Caution: For educational use only.