NeuLog Motion Logger Sensor
Supplier: Wards
NUL213
470137-394EA 165.05 USD
470137-394
NeuLog Motion Logger Sensor
Data Loggers
This sensor uses an ultrasonic transducer to provide streamlined measurements in the classroom.
  • Sensor measures echo return to determine distances
  • Three modes of operation: Distance, velocity, and acceleration
It supports experiments lasting from 1 sec to 31 days.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
Caution: For educational use only.