NeuLog Pressure Logger Sensor
Supplier: Wards
NUL210
470137-388EA 142.7 USD
470137-388
NeuLog Pressure Logger Sensor
Data Loggers
The NeuLog pressure logger sensor helps you monitor chemical reactions involving gases.
  • Applicable to Boyle's law and Gay-Lussac's law
  • Ideal sensor for studying weather phenomena
  • Sensor monitors chemical reactions involving gases
  • For experiments between one second and 31 days long
  • Maximum sample rate of 100 S/s
It also helps you investigate Boyle's law and Gay-Lussac's law for ideal gases. It's an perfect sensor for studying weather phenomena.
The sensor is connected to a small, stainless steel tube that connects to pressure sources.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
Caution: For educational use only.