National 200 Series Compound Microscope
Supplier: National Optical
215-RLED
470136-760EA 831 USD
470136-760
National 200 Series Compound Microscope
Microscopes
These affordable microscopes include a number of high-quality features.
  • Wide-Field 10x Eyepiece with Pointer
  • Bulb Life: 10,000 Hours
  • Four-Hole Reversed Objective Turret
Features include a wide-field 10x eyepiece with pointer and a four-hole reversed objective turret.