Acid Rain and the Environment: Acidity and Plant Growth
Supplier: Ward's Science
IS9704
470136-736KT 88.8 USD
470136-736
Acid Rain and the Environment: Acidity and Plant Growth
Educational Classroom Kits and Activities
Show How Acid Rain Affects Plants.
  • Real world hands on science
  • Ideal for biology, chemistry and earth science classrooms!
The problem of acid rain is quite often associated with its effects on aquatic systems. But it can also have devastating effects on terrestrial environments. In this activity, students will examine the detrimental effects of acidic conditions on plants grown under normal soil conditions, mildly acidic soil conditions, and very acidic soil conditions. Through physical observation, students will determine if the acidity has any impact on the growth of the plant.
Ordering information: Includes Hydrochloric Acid, Potting Soil, Transulcent Cups, Absorbent Wick Pad, Radish Seeds, and Teacher's Guide and Student Copymasters.