Green Nanochemistry: Synthesis of Silver Nanoparticles
Supplier: Ward's Science
IS9506
470136-726KT 68.5 USD
470136-726
Green Nanochemistry: Synthesis of Silver Nanoparticles
Educational Materials Chemistry Educational Materials Green Chemistry Learning Activities
Synthesize Silver Nanoparticles from Silver Nitrate.
  • Green chemistry inquiry activity
  • Non-hazardous alternative chemicals
  • All chemicals required are included
Certain materials, when prepared at nanoscale levels, display different physical and chemical properties than those of the same materials prepared on a larger scale. In this activity, students synthesize silver nanoparticles from silver nitrate. The particles are synthesized in glucose and starch, as opposed to the more hazardous reagents typically used.
Includes:
Silver Nitrate
Dextrose
Soluble Starch
Sodium Hydroxide
Teacher's Guide and Student Copymasters