Detergents and the Environment
Supplier: Ward's Science
IS9502
470136-718KT 55.9 USD
470136-718
Detergents and the Environment
Educational Materials Chemistry Educational Materials Green Chemistry Learning Activities
Learn How Detergents Affect Aquatic Environments.
  • Green chemistry
  • Ideal for chemistry, biochemistry, environmental and earth science courses
  • All chemicals required are included
This complete kit helps students understand the consequences of cultural nutrient overloading on the aquatic environment. In your classroom, students can monitor the effects of two detergent builders on natural water samples, as well as visually quantify the differences between phosphate and non-phosphate detergent builders on aquatic organisms. They will also learn about more environmentally friendly alternatives in typical consumer chemical goods.
Ordering information: Includes Phosphate Buffer Solution, Non-Phosphate Buffer Solution, Control (Deionized) Water, Plastic Cups, Microscope Slides, Coverslips, Teacher's Guide and Student Copymasters.