Celestron Telescope Eyepiece and Filter Kit, 1.25"
Supplier: Celestron International
94303
470136-650EA 299.19 USD
470136-650
Celestron Telescope Eyepiece and Filter Kit, 1.25"
Telescopes
This 1.25" eyepiece and filter kit includes 14 accessories that will add new life to your telescope and your observing!
  • The most popular planetary filters are included in this kit
Have fun seeing what a blue filter will reveal about jupiter or unveil the polar ice caps on mars with a red or orange filter. A moon filter will increase your enjoyment of lunar observations by dimming the brightness and increasing contrast.
Delivery information: You’ll get five quality Plossl eyepieces, a 2X Barlow Lens, six colored filters to improve observations of the planets, a Moon filter, and a foam-fitted aluminum carry case with extra room to grow. Five eyepieces and a Barlow lens equals 10 different magnifications to choose from, including the equivalent of a 3, 4, 6, 6.5, 8, 8.5, 13, 16, 17 and 32 mm. And that doesn’t include eyepieces you may already have in your collection, which will now achieve double the magnification with the addition of the Barlow.