Laboratory Refrigerators and Freezers, Thermo Scientific
Supplier: Thermo Fisher Scientific
02LFEETSA
470136-644EA 1399 USD
470136-644
Laboratory Refrigerators and Freezers, Thermo Scientific
Refrigerators
These refrigerators, freezers, and combination units provide a safe and economical solution for sample storage. Suited for routine laboratory applications. Store samples safely and economically
  • All units feature environmentally friendly and non-CFC refrigerants
  • Adjustable temperature controls
  • Easy-to-clean and has a durable finish
  • All freezers are manually defrost
Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature excellent temperature stability.
Certifications: cCSAus.
Caution: Note: These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up, and temperature performance features. These units are not suitable for corrosive environments.