Semicircular Refraction Cell - Acrylic
Supplier: United Scientific Supplies
RCSC01
470136-570EA 6.5 USD
470136-570
Semicircular Refraction Cell - Acrylic
Culture Dishes Petri Dishes
Investigate the optical properties of liquids.
  • Demonstrate internal reflection and refraction
  • Semicircular design allows for each light ray tracing
These clear acrylic refraction cells are excellent for showing refraction and internal reflection in water, and to measure its refractive index.  The cells have uniform wall thickness to eliminate distortions. 12 cm diameter x 2.5 cm high.