Formalin Neutralizing Pads, 11"×10"
Supplier: Azer Scientific Inc.
ES728
470136-436CS 395.4 USD
470136-436
Formalin Neutralizing Pads, 11"×10"
Trays/Dishes/Pots Dissecting Trays
Create a safer and more enjoyable dissection experience with these odor-absorbing pads, designed to minimize exposure to formaldehyde off-gassing and neutralize excess formalin.
  • Removes odor
  • Decrease distraction
Create a safer and more enjoyable dissection experience with these odor-absorbent pads, designed to minimize exposure to formaldehyde off-gassing and neutralize excess formalin. Simply line your specimen pan with a pad and place the specimen on top to immediately absorb excess formalin and odors during specimen dissection.