Eisco® Dicot Stem Anatomy
Supplier: Wards
BM0010
470136-376EA 233.7 USD
470136-376
Eisco® Dicot Stem Anatomy
Models Botany Models
Model shows the transverse and longitudinal section of a dicotyledonous stem in which case the cambium ring has been formed but no secondary growth has yet taken place.
Very useful model for study of epidermis, cork layer, cork cambium, cortical parenchyma, starch sheath medullary rays, phloem, sieve plate, sieve tube, phloem parenchyma and inter fasicular cambium, xylem, pitted vessels pith and more.