Eisco® Endocrine System Model
Supplier: Wards
AM0346
470136-314EA 111.05 USD
470136-314
Eisco® Endocrine System Model
Models Human Biology Models
Shows position and structure of major glands.
  • Shows position and structure of major glands
  • Includes pineal, thyroid, thymus, heart, adrenals, kidneys, pancreas and pancreatic duct, ovaries, testes, and placental hormones
  • Hand-painted by skilled artists to deliver the highest level of detail possible
A frontal section of the human body, in miniature, shows major glands of the endocrine system. Glands and organs in the model are larger than scale to allow for easier viewing. Both glands of male and female are present.