Ward's® Graduated Mechanical Stage, Motic
Supplier: VWR
MSH-008
470136-220EA 70.8 USD
470136-220
Ward's® Graduated Mechanical Stage, Motic
Microscope Stages
Upgrade your plain-stage microscope.
  • Easily attaches to pre-drilled plain stages
  • Stage accommodates all slide sizes
  • Fits all of WARD'S compound microscopes
Precisely position slides with this graduated mechanical stage, which easily attaches to pre-drilled plain stages. Finely calibrated in 1 mm increments, the stage accommodates all slide sizes. It is constructed for right-hand access to both adjusting knobs and fits all of Ward's® compound microscopes.