Eisco® Premium Enlarged Human Ear Model, 5 Parts
Supplier: Wards
AM0042
470136-040EA 171.45 USD
470136-040
Eisco® Premium Enlarged Human Ear Model, 5 Parts
Models Human Biology Models
Multipiece enlarged model for advanced anatomical studies of the human ear.
  • Enlarged (5x) human ear model
  • Includes 5 removable piece
  • Mounted on base
  • Hand painted
  • Included numbered key card
This hand painted human ear model allows detailed study of the outer, middle, and inner ear. Five removable parts include: petros portion of the temporal bone section of the auditory canal, labyrinth, tympanic membrane with malleus, and incus.