Digital Orbital Shaker
Supplier: Edvotek
OR-100-12
470134-648EA 1399 USD
470134-648
Digital Orbital Shaker
Shakers and Mixers Orbital Shakers
Variable Speed for Orbital Motion Study
  • Rugged, washable sample pad
  • Suction cup feet prevent "walk away"
  • Features a full one-year warranty
  • 1 year warranty
For experiments requiring orbital motion, this variable speed model is the ideal choice. Select either continuous or a timed run mode. Includes a digital timer that can be set for as little as 1 min. up to 99 hrs. 59 min.