O₂ Gas Sensor to Spirometer Adapter
Supplier: Vernier Software & Technology
O2-SPR
470133-506EA 7.99 USD
470133-506
O₂ Gas Sensor to Spirometer Adapter
Fittings and Connectors
This adapter facilitates the connection between an Oxygen Gas Sensor and a Spirometer. Measures the oxygen gas concentration and flow rate of exhaled air.