MyBlock Mini Dry Bath with Heated Lid
Supplier: Ward's
BSH200-HL
470133-476EA 577.4 USD
470133-476
MyBlock Mini Dry Bath with Heated Lid
Block Heaters Digital Block Heaters
Heat your samples in minutes
  • Heated lid prevents evaporation and condensation
  • Compact, fits in the palm of your hand
  • Independent temperature control of lid and block
  • Room temperature to 37°C in just 2 minutes
  • Blocks sold separately
The new MyBlock™ HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation.Both the block and lid temperature can be selected independently in increments of 0.1°C, making this MyBlock the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions & other temperature hyper-sensitive procedures.
Ordering information: Two year warranty.