TeacherGeek Bulk motors
Supplier: TEACHERGEEK INC
1823-75
470132-511EA 3.09 USD
470132-511
TeacherGeek Bulk motors
Educational Materials Physics Educational Materials TeacherGeek System Components
Extra motors used in our popular Catch the Bug Activity.
  • 35.06mm length including the shaft
  • Shaft by itself is 5.76mm long and 2mm in diameter
  • 1–3 volts