TeacherGeek Check Valve - ¹/₈"
Supplier: TEACHERGEEK INC
1823-57
470132-476EA 12.96 USD
470132-476
TeacherGeek Check Valve - ¹/₈"
Educational Materials Physics Educational Materials TeacherGeek System Components
Check valves allow one way passage of water or air through tubing.
They are used in pumps, jacks and other hydraulic/pneumatic applications.