TeacherGeek Gear Set - 40-PIECE mixed colors
Supplier: TEACHERGEEK INC
1823-28
470132-424EA 19.98 USD
470132-424
TeacherGeek Gear Set - 40-PIECE mixed colors
Educational Materials Physics Educational Materials TeacherGeek System Components
Gear set to expand your TeacherGeek ideas.
The TeacherGeek 40 Piece Multi-Color Gear Set contains ten each of the following sizes: 50 tooth, 40 tooth, 20 tooth, 10 tooth. These are used to create gear transmissions such as for vehicles, wind turbines, lifts, pumps or whatever you wish to gear up or down for the desired output. The splined holes grip the dowel axle for a secure non-slip fit.
Multi-Color Set includes:
•10 - 10 tooth gears
•10 - 20 tooth gears
•10 - 40 tooth gears
•10 - 50 tooth gears