Dual Output Electrophoresis Power Supply
Supplier: VWR International
470130-692
470130-692EA 183 USD
470130-692
Dual Output Electrophoresis Power Supply
Power Supplies
Double the Voltage
Featuring two different voltages, 70 V or 125 V, this dual output power supply is flexible. The output voltage is easily selected via a rocker switch and is available on two parallel pairs of safety sockets. The unit’s size is 7 × 5.5 × 4" (18 × 14 × 10 cm).