BRIGHT™ Science Kit
Supplier: BRIGHT OF SWEDEN
102-1
470129-342EA 442.99 USD
470129-342
BRIGHT™ Science Kit
Models Molecular Biology Models
Complete Sets for Students and Teacher
  • Hands on activity to help students learn
  • Teach chemical bonding and molecular associations in an interactive form
  • Mix and match components to teach various concepts
Give every student a chance to get their hands on these innovative magnetic atom parts, with eight student atoms and one teacher model, along with suggested lesson plans and student worksheets. These innovative pieces provide students with a concrete manipulative tool for exploring abstract concepts of atomic structure, isotopes and ion formation. Large, simple parts are easy to identify and clearly show structures for easy understanding.
Caution: WARNING: Choking Hazard. Small Parts. Not for Children under 3 yrs.