Heavyweight Spill Control Pads
Supplier: ICC THE COMPLIANCE CENTER INC.
AB-YPB50H
470129-054PK 115.3 USD
470129-054
Heavyweight Spill Control Pads
Spill Control Materials
Quickly Absorbs Chemicals and Liquids
When hazardous or unknown liquids spill, use these heavyweight spill pads for quick, effective response. Spill pads are yellow for quick identification in an emergency. They are made of 100% polypropylene and absorb the most aggressive and caustic fluids. Sonic–bonded “dimples” add strength and fast wicking power. Box of 50.