Biotic Indicators Riker Mount
Supplier: WHITE OWL ENTERPRISES
R287
470128-618EA 104.95 USD
470128-618
Biotic Indicators Riker Mount
Specimens Preserved Specimens Display Specimens
Riker mount of biotic indicators for display.
  • High quality display mount
This comprehensive display is a great tool for reviewing water quality indicators or identifying aquatic macro-organisms from field studies. Featured first are the 'EPT' larval specimens (Ephemeroptera, Plecoptera, Trichoptera) which are very sensitive to pollution and turbidity. Next are seven species commonly found in healthy water systems and tolerant of moderate pollutants. Finally, the 'tolerant' species are represented by four specimens including a lunged snail and leech. Each specimen is fully labeled.