Battery Operated Light Box
Supplier: Wards
PH0600-LED
470128-434EA 39.3 USD
470128-434
Battery Operated Light Box
Lights Light Boxes
This power–saving alternative to a traditional ray box is portable and durable.
  • Led Prevents Heat Buildup
  • Battery-Operated for Portability
  • Sturdy Metal Housing
The low-power LED mounted inside this metal ray box prevents heat buildup for added safety over extended periods.
The unit operates on two AA batteries and is fitted with two 4 mm sockets for the battery pack, which is easily mountable on the ray box.
Ordering information: This box includes a collimating lens and slits with accommodating slots in integral side rails. Two AA batteries are required but not included.