Variable Inductor
Supplier: VWR International
PH0845VIC
470128-394EA 374.5 USD
470128-394
Variable Inductor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Adjustable for Precision
  • Compatible with 4 mm safety sockets.
  • Includes a carrying handle for easy portability. 
The screw adjustment enameled wire winding built into this unit allows precise measurements up to 1.3 H. Soft iron core displacement includes a scale mark and quick advancement worm.