Acid Spill Clean Up Kit
Supplier: Ward's Science
IS5003
470127-676EA 74.8 USD
470127-676
Acid Spill Clean Up Kit
Spill Control Materials
Ready to Use for Quick Response.
  • Handy compact design
  • Contains everything to safely contain the spill
Spills are inevitable in almost every chemistry lab. Be prepared for potentially dangerous acid spills with this complete kit that includes everything you need for safe cleanup and disposal. Includes one bag of Vermiculite absorbent, one bag of neutralizing mixture, two vinyl exam gloves, two 9" x 16"polybags, two blank shipping tags, twist ties, and a dust pan and brush set. A detailed instruction sheet/SDS is provided.