Caustic Spill Clean Up Kit
Supplier: Ward's Science
IS5002
470127-674EA 75.1 USD
470127-674
Caustic Spill Clean Up Kit
Spill Control Materials
Safe and Easy Clean-Up.
  • Handy compact design
  • Contains everything to safely contain the spill
Safely respond to accidental caustic spills in your chemistry lab with this complete, ready-to-use kit. Includes one bag of Vermiculite absorbent, one bag of neutralizing mixture, two 9" x 16" polybags, two blank shipping tags, two vinyl exam gloves, twist ties, and a dust pan and brush set. A detailed instruction sheet/SDS is provided.