Solvent Spill Clean Up Kit
Supplier: Ward's Science
IS5001
470127-672EA 70.5 USD
470127-672
Solvent Spill Clean Up Kit
Spill Control Materials
For Safe Disposal of Chemical Spills.
  • Handy compact design
  • Contains everything to safely contain the spill
Keep this kit on hand in your chemical storeroom or laboratory to be prepared for safe clean-up of inevitable solvent spills. Includes one bag of diatomaceous earth, two vinyl exam gloves, two 9" x 16" polybags, two blank shipping tags, twist ties, and a dust pan and brush set. A detailed instruction sheet/SDS is provided within the kit.