GPI Anatomicals® Clear Ear Model
Supplier: GPI ANATOMICALS
2251
470126-650EA 101.6 USD
470126-650
GPI Anatomicals® Clear Ear Model
Models Human Biology Models
Reveals Middle And Inner Structures
This life-sized human ear model is transparent to allow easy visual demonstration and explanation of basic inner-workings of the ear and conditions such as ear infections, tinnitus, hearing disorders and deafness. Middle and inner-ear components that are usually hidden are clearly revealed, including the tympanic membrane, auditory ossicles, semicircular canals, and the Cochlea. The malleus, incus, and stapes bones that compose the auditory ossicles can also be seen in full detail.