Rescue Critters® K-9 Intubation Trainer
Supplier: THALES & CO.
606
470126-192EA 998.89 USD
470126-192
Rescue Critters® K-9 Intubation Trainer
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Animal Intubation Practice
  • Head only trainer for compact practice
  • Airway and working lungs
  • Made to order
Elegantly simple – this model is built for performing proper endotracheal placement. The ET tube either goes correctly into the trachea or wrongly into the esophagus. Improved representations of the trachea, esophagus and epiglottis are all realistically crafted into this dog bust. It also features an airway with working lungs. Because the head is attached to a table clamp with a pivot base, it can be easily manipulated.
Ordering information: This item is made to order. Returns cannot be accepted.