AC/DC power supply
Supplier: Wards
EPR1329A
470125-556EA 270 USD
470125-556
AC/DC power supply
Power Supplies
Ideal for charging wet cell batteries, electroplating, electrolysis, and many other applications.
Output is electronically protected and the line fuse protects against internal shorts. A front panel voltage knob makes adjustments easy and color-coded 4mm banana jacks provide output. Includes a three-wire grounded cord and plug.
Certifications: CSA Approved