LevGo® smartSpatula® Disposable Lab Spatula
Supplier: Avantor
470125-222PK 6.8 USD
470125-222
LevGo® smartSpatula® Disposable Lab Spatula
Scoops
Great for handling small amount of materials.
  • Utilitarian size: 210×7 mm (8.5×0.25"), 20/pack
  • Spoon/scraper fits into 15 ml tube and scoop fits into 0.5 ml tube
  • 100% polypropylene is autoclavable and resistant to dilute acids and bases
  • Blue contrasts with white material
Versatile double-ended polypropylene tool that can scoop, scrape, spoon, stir, collect, transfer, handle or manipulate a variety of chemicals and biological materials.
Ordering information:
The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc., patent 8,709,362.