SP Bel-Art Wash Bottle Carrier, Bel-Art Products, a part of SP
Supplier: Bel-Art
F11640-0001_SE
470123-962EA 83.15 USD
470123-962
SP Bel-Art Wash Bottle Carrier, Bel-Art Products, a part of SP
Racks Bottle Racks
Safely and conveniently transport up to 6 wash bottles in this sturdy, lightweight polypropylene carrier. Its secure handle affords both comfort and control. Autoclavable at 121 °C (250°F) and chemical resistant. Holds 250 ml (8.4oz.) or 500 ml (16.9oz.) bottles. Each hole diameter is 7.4 cm (2.91").