Washing Bottle with Trigger Sprayer
Supplier: Tolco
120112110412
470123-944EA 11.15 USD
470123-944
Washing Bottle with Trigger Sprayer
Bottles Wash Bottles
Great for general lab use.
  • Round polyethylene bottle with threaded neck
  • Polypropylene screw-on, trigger-style sprayer
  • Knurled nozzle adjustable from fine mist to jet stream