Dropping Bottle Block
Supplier: Bennett Wood Specialties
6793 + PKG
470123-936EA 38.35 USD
470123-936
Dropping Bottle Block
Racks Bottle Racks
Holds six 30 mL round dropping bottles. Oil treated wood block with rubber feet, drilled with holes 1-1/2" dia. Size: 12" W x 2-1/4" D. Without bottles.