Folding Digital Thermometer
Supplier: General Tools Manufacturing
PT300M
470123-712EA 80 USD
470123-712
Folding Digital Thermometer
Thermometers Digital Thermometers
Large, Easy to Read Digital Display.
  • Measures in both C° and F°
  • Display turns on automatically when the probe is unfolded
  • Average response time 1 second
Stores the minimum and maximum temperatures.
Ordering information: Meter includes one AAA battery and protective sleeve.