VWR® Pipette Fillers
Supplier: Heathrow Scientific
HS20632A
470123-222CS 2181.9 USD
470123-222
VWR® Pipette Fillers
Pipettes Pipette Bulbs
Pipette fillers eliminate the hazardous practice of drawing liquids into pipettes by mouth.
  • Filters work with pipettes up to 50 ml in size
  • Filters help avoid sample contamination
  • Safe, precise delivery
  • Ideal for volatile, corrosive, or poisonous liquids
  • Fits a standard serological pipet
To use, insert the pipette into the bottom opening of the filler, draw to the desired volume, and expel by pressing lettered valves.
The filters are made of soft rubber. Colors may vary. Dimensions: 54×54×130 mm (L×W×D).