Variable Volume Economy Micropipettes
Supplier: United Scientific Supplies
PVV-10 PVV-100 PVV-200 PVV-1000
470123-180EA 3.67 USD
470123-180 470123-182 470123-184 470123-186 470200-654
Variable Volume Economy Micropipettes
Pipettes Micropipettes
High performance pipette.
  • Compatible with Gilson and eppendorf brand tips
  • Built-in tip ejector
  • Click stop digital system
  • Easy calibration and maintenance
  • Autoclavable tipcone assembly
Easy to disassemble for servicing and autoclaving. The 100 to 1000 μl micropipette uses 1000 μl tips. All other micropipettes use standard 200 μl tips.