5 MHz Function Generator with Display
Supplier: B & K Precision
4011A
470123-168EA 659.19 USD
470123-168
5 MHz Function Generator with Display
Batteries
Outstanding general-purpose unit.
  • Frequencies from as low as 0.5Hz, up to 5MHz, in 7 ranges
  • Accuracy of ±10ppm, ±1 count
  • 4-digit LED display
  • Variable amplitude output (max 10Vp-p into 50) with coarse and fine adjustment
  • VCG input, variable DC offset and duty cycle
  • 20dB step attenuator
  • 50 ±10% output impedance
This generator produces sine, triangle, square pulse and range signals, and has a separate TTL/CMOS output. It comes with an output cable (BNC/insulated clips), a power cord, and an instruction manual.