Cobalt Chloride Test Paper
Supplier: Precision Laboratories
150-12V-100
470123-112PK 28.7 USD
470123-112
Cobalt Chloride Test Paper
Test Paper Indicator Paper
Cobalt Chloride cloth detects the presence of moisture to determine if you have wetness or dampness problems.
The Cobalt chloride cloth is responsive to 50% relative humidity and above, and any direct measure of an aqueous solution.
Packaging: 12 Vials of 100 Strips.