Basic Molecular Individual Model Kit
Supplier: Lab-Aids
125
470122-966KT 50.35 USD
470122-966
Basic Molecular Individual Model Kit
Models Molecular Biology Models
An Ideal Building Set for Creating Basic Molecular Structures of Many Chemical Compounds
  • Requires one 50 minute class period to complete
  • Reusable materials
This basic molecular model kit is ideal for teacher demonstrations or individual student activities. Construct ten different molecular models, including methane, water, ammonia, hydrogen sulfide, ethane, and propane. The kit contains 191 components plus a self-pace instruction book.
Ordering information: Kit includes Covalent electron bond; Carbon tetrahedral (black), centers; Nitrogen tetrahedral (red), centers; Oxygen tetrahedral (blue), centers; Sulfur tetrahedral (yellow), centers; Chlorine single electron (green), atoms; Hydrogen single electron (white), atoms; Oxygen double electron (blue), atoms; Sulfur double electron (yellow), atoms, and Student Directions and Guide.