Classroom PCR LabStation
Supplier: Edvotek
5067
470122-884EA 5999 USD
470122-884
Classroom PCR LabStation
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
A complete package for your classroom PCR needs.
  • Supports up to 25 students
  • Includes all materials necessary to perform PCR assays start to finish
Kit includes: 1× EdvoCycler™ 2 (48×0.2 ml); 6× M12 Complete™ package (14×7 cm tray and 7×7 cm trays (2)); 3 x model 509 DuoSource™ Power Supplies (100 or 150 V, for 1 to 2 units); 6× Variable MicroPipette (5 to 50 µl); 1× TruBlu™ 2 LED transilluminator and 1× 3 L water bath.