Polypropylene Beaker Set
Supplier: United Scientific Supplies
BPSET5-SW
470122-772KT 32.9 USD
470122-772
Polypropylene Beaker Set
Beakers
Made of unbreakable polypropylene, this beaker set avoids dangerous accidents.
  • Beakers Can Withstand up to 120°C
  • Autoclavable
  • Permanent Graduations
  • Unbreakable
  • No-Drip Pouring Spouts
The beakers are ideal for educational use and feature permanent graduations and no-drip pouring spouts.
They are autoclavable and can withstand temperatures up to 120°C.
Ordering information: These beakers come in a polyethylene bag.