Beaker Starter Set
Supplier: United Scientific Supplies
BGSET5
470122-762CS 69.5 USD
470122-762
Beaker Starter Set
Beakers
An ideal starter set for any laboratory.
  • Feature pour spouts
  • Double-graduated metric scale and durable marking area
A set of 5 beakers of the most commonly used sizes.
Delivery information: Contains one each of the 50, 100, 250, 600, and 1000 ml beakers.