Straight Filament Lamp Bulbs
Supplier: Grainger
4V461
470110-278EA 15.55 USD
470110-278 470122-618
Straight Filament Lamp Bulbs
Lights Incandescent Lights, Lamps and Bulbs
Single-line filament is ideal for student diffraction experiments. It's also a great way for students to observe filament technology in action, because the slender 15.2 cm lamp lets them clearly and distinctly see the entire process.
Medium screw base and 120 V, suitable for standard sockets.