CPEP Fusion, Physics of Fundamental Energy Charts
Supplier: CONTEMPORARY PHYSICS EDUCATION
397341
470122-544EA 13.8 USD
470122-544
CPEP Fusion, Physics of Fundamental Energy Charts
Charts
Visualize fusion and energy.
  • Students will be able to compare the energy released in a variety of conversion processes, including combustion, fission and fusion
Developed by the Fusion and Plasmas Group of the Contemporary Physics Education Project (CPEP), Fusion presents up-to-date information on plasma physics and fusion reactions, examples of solar and terrestrial fusion reactions, explains how Einstein’s famous E=mc2 describes the process of fusion and offers illustrations of everyday and exotic plasmas.